Learn More
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Brand: Corning™ 4413
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.

Specifications
Strip | |
Autoclavable | |
8-Strip Microplate Cluster Tube | |
12/Pk. | |
Translucent | |
Rack | |
0.033 oz. |
12/Rack | |
Polypropylene | |
Sterile | |
120/Cs. | |
1 mL | |
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
96 |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.