Learn More
Epredia™ Cover Slips, Rectangles
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes and thicknesses.
Brand: Epredia™ A67761330
This item is not returnable.
View return policy
Description
- Highly transparent and achromatic – practically free of any color distortion
- Good planarity
- Refraction index adjusted for microscopes
- High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
- Very good chemical stability
Specifications
Rectangle | |
48 mm | |
64 x 48 mm |
64 mm | |
#1 (0.13 to 0.16 mm) | |
100/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Epredia™ Cover Slips, Rectangles >
Spot an opportunity for improvement?Share a Content Correction