Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall
GREENER_CHOICE
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
£5.55 - £5.80
Specifications
Product Type Hooded Coveralls
Material Polyethylene
Color White
Ankle Style Elastic
Neck Style Hooded
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15310541
GREENER_CHOICE
View Documents
DuPont™
D14886039
Small
£5.55
Each
In Stock
to see stock.
Edge
Add to basket
15320541
GREENER_CHOICE
View Documents
DuPont™
D14886047
Medium
£5.55
Each
In Stock
to see stock.
Edge
Add to basket
15330541
GREENER_CHOICE
View Documents Promotion Details
DuPont™
D14886050
Large
£5.55
Each
In Stock
to see stock.
Edge
Add to basket
15360541
GREENER_CHOICE
View Documents
DuPont™
D14886081
3X-Large
£5.80
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
15350541
GREENER_CHOICE
View Documents
DuPont™
D14886075
2X-Large
£5.55
Each
In Stock
to see stock.
Edge
Add to basket
15340541
GREENER_CHOICE
View Documents Promotion Details
DuPont™
D14886064
X-Large
£5.55
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
Description
Description
  • Robust yet lightweight
  • Elasticated face, wrists, waist and ankles
  • Antistatic treatment on the outside
  • Stitched external seams in yellow for visual identification and differentiation
Specifications
Specifications
Hooded Coveralls
White
Hooded
Zipper
Category III, Type 5 and 6
Polyethylene
Elastic
2-panel
Stitched
(EU) 2016/425, EN 1073-2, EN 1149-1
Videos
SDS
Documentos
Documentos
Product Certifications
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado