Learn More
Greiner Bio-One Cryo.s Cryogenic Tubes with Internal Thread
Manufactured with high-grade polypropylene for strength and high thermal resistance. Greiner BioOne™ Cryo.s Cryogenic Tubes with Internal Threads withstand a temperature range of -196 to +121°C.
£94.91 - £158.17
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Polypropylene |
Temperature Range (Metric) | -196°C to +121°C |
Sterility | Sterile |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Closure Color | Bottom Shape | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Color | Bottom Shape | Includes | Price | Quantity & Availability | ||
10627671
|
Greiner Bio-One
123279 |
1 mL | Blue | Conical Bottom, Starfoot Base | Writing Area, Cap Inserts |
£126.54
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10401721
|
Greiner Bio-One
123277 |
1 mL | Green | Conical Bottom, Starfoot Base | Writing Area, Cap Inserts |
£126.54
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10617671
|
Greiner Bio-One
123278 |
1 mL | Yellow | Round | Writing Area, Cap Inserts |
£126.54
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10329910
|
Greiner Bio-One
121279 |
2 mL | Blue | Round | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10686321
|
Greiner Bio-One
122277 |
2 mL | Green | Round (Starfoot) | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10175970
|
Greiner Bio-One
121280 |
2 mL | Red | Round | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10147480
|
Greiner Bio-One
122280 |
2 mL | Red | Round (Starfoot) | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10563821
|
Greiner Bio-One
121278 |
2 mL | Yellow | Round (Self-standing) | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10450181
|
Greiner Bio-One
124274 |
5 mL | Blue | Round | Writing Area, Cap Inserts |
£94.91
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10459990
|
Greiner Bio-One
124273 |
5 mL | Red | Round | Writing Area, Cap Inserts |
£94.91
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10175350
|
Greiner Bio-One
124263 |
5 mL | Natural | Round | Writing Area, Cap Inserts |
£94.91
Pack of 300 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10063950
|
Greiner Bio-One
122278 |
2 mL | Yellow | Round (Starfoot) | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10701391
|
Greiner Bio-One
122263 |
2 mL | Natural | Round Bottom, Starfoot Base | White writing area; cap inserts for labeling |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10731201
|
Greiner Bio-One
121277 |
2 mL | Yellow | Round | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10033990
|
Greiner Bio-One
122279 |
2 mL | Blue | Round (Starfoot) | Writing Area, Cap Inserts |
£158.17
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10462481
|
Greiner Bio-One
123280 |
1 mL | Red | Conical Bottom, Starfoot Base | Writing Area, Cap Inserts |
£126.54
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Free of detectable human DNA, endotoxins, DNase and RNase
- Sterile, non-pyrogenic, non-cytotoxic, no heavy metals contained
- CE-marked (CE/IVD)
- As a safety precaution Cryo.s should be stored in a freezer or exclusively in the gas phase over the liquid nitrogen
- Suitable for shipment by air
- With white, scratch-resistant writing area
- 150 inserts per case (for additional labelling on the cap)
- Suitable for storage at -20°C, -80°C and in the vapour phase of LN2
- Multiple cap colours and working volumes
- Ultra-pure medical grade, USP class VI certified polypropylene w/o leachables
- Accessory storages boxes and working racks available
Specifications
Not Autoclavable | |
-196°C to +121°C | |
Yes |
Polypropylene | |
Sterile | |
Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Greiner Bio-One Cryo.s Cryogenic Tubes with Internal Thread