missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CTS™ KnockOut™ SR XenoFree Kit
CTS™ KnockOut™ SR XenoFree Kit
£114.00 - £434.00
Specifications
Culture Environment | CO2 |
---|---|
Cell Type | Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) |
Classification | Serum-free, Xeno-free |
Form | Frozen |
Product Type | Stem Cell Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10195684
|
Gibco™
A1099201 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10577024
|
Gibco™
A1099202 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
GIBCO™ KnockOut™ SR XenoFree CTS™ enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, which facilitates the transition of hESC and hiPSC research from the bench to the clinic.The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
GIBCO™ KnockOut™ SR XenoFree CTS™ does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, KnockOut™ SR XenoFree CTS™ can be used for hESC⁄hiPSC cryopreservation, derivation, and differentiation studies.
GIBCO™ CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
The KnockOut™ SR XenoFree CTS™ Kit includes a bottle of KnockOut™ SR XenoFree CTS™ as a frozen liquid in a 500 mL or a 100 mL PET bottle and a product manual.
This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.
Specifications
CO2 | |
Serum-free, Xeno-free | |
Stem Cell Media | |
Sterile | |
No Glucose, No Glutamine, No HEPES, No Phenol Red | |
CTS™, KnockOut™ | |
Human |
Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) | |
Frozen | |
Serum-free | |
HSA, Sodium Bicarbonate | |
≤ 10 EU/mL | |
ES Cell-Qualified |
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.