missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Cubis™ II Balance

The Cubis™ II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a range of accessories allows the individual adaptation to all weighing tasks.
£9145.00 - £25210.00
Product Code | Brand | Capacity | Model | Readability | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Model | Readability | Price | Quantity & Availability | |||
30052882
![]() |
Sartorius
MCA125P-I-MTS |
60 g, 120 g | MCA with Draft Shield I | 0.01 mg, 0.1 mg |
£15735.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052877
![]() |
Sartorius
MCA125P-U-MTS |
60 g, 120 g | MCA with Draft Shield U | 0.01 mg, 0.1 mg |
£12075.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052889
![]() |
Sartorius
MCA125S-I-3-MTS |
120 g | MCA with Draft Shield D | 0.01 mg |
£16950.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052881
![]() |
Sartorius
MCA225S-I-3-MTS |
220 g | MCA with Draft Shield D | 0.01 mg |
£22035.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052883
![]() |
Sartorius
MCA225S-I-MTS |
220 g | MCA with Draft Shield I | 0.01 mg |
£22070.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052878
![]() |
Sartorius
MCA225S-U-MTS |
220 g | MCA with Draft Shield U | 0.01 mg |
£18405.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052886
![]() |
Sartorius
MCA226S-I-3-MTS |
220 g | MCA with Draft Shield D | 0.005 mg |
£25210.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052869
![]() |
Sartorius
MCE125P-U-MTS |
60 g, 120 g | MCE with Draft Shield U | 0.01 mg, 0.1 mg |
£9145.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052876
![]() |
Sartorius
MCE125S-I-3-MTS |
120 g | MCE with Draft Shield D | 0.01 mg |
£14020.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052879
![]() |
Sartorius
MCE125S-U-MTS |
120 g | MCE with Draft Shield U | 0.01 mg |
£10135.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
30052888
![]() |
Sartorius
MCE225S-I-3-MTS |
220 g | MCE with Draft Shield D | 0.01 mg |
£19105.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Leading performance. Monolithical weighing system, integrated climate sensors, individual sample holders Error-free operation. Individual QApp workflows, motorized auto-leveling
- Full end-to-end data integrity. 21 CFR Part 11 compli-ance, integrated audit-trail, state-of-the-art user management
- Outstanding service support. Integrated status center, integrated service functions preventive maintenance on accredited standards
Specifications
Color Touchscreen | |
Balance | |
Class I |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius Cubis™ II Balance