Learn More
Thermo Scientific™ Culti-Loops™ Trichophyton equinum ATCC™ 12544™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4608302
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
12544TM* | |
Trichophyton equinum | |
Culti-Loops Quality Control Organism | |
Qualitative QC | |
5 Loops/Pk. |
Culti-Loops are CE Marked. | |
Culti-Loops | |
Fungi | |
5 Loops/Pack | |
CDC B-56 |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
155868 | Certificate of Analysis | 16/09/2024 | R4608302 |
129057 | Certificate of Analysis | 10/07/2024 | R4608302 |
237836 | Certificate of Analysis | 16/04/2024 | R4608302 |
526488 | Certificate of Analysis | 04/04/2024 | R4608302 |
670049 | Certificate of Analysis | 03/04/2024 | R4608302 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.