Learn More
R&D Systems™ Cultrex Poly-L-Ornithine Solution
R&D Systems™ Cultrex™ Poly-L-Ornithine Solution is a highly positively charged amino acid chain, which is generally used as a coating reagent to promote cell adhesion in culture.
Brand:  R&D Systems™ 3436-100-01
Product Code. 18777696
  • £141.00 / 100mL
Explore more special offers
Description
Description
  • Poly-L-Ornithine along or in combination with Fibronectin or Laminin-I is frequently used to enhance attachment and differentiation of various types of neuronal and neural stem cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
Product is stable for at least 6 months from the date of receipt when stored at 2 - 8°C. Keep sterile.
Multi-species
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Cultrex Poly-L-Ornithine Solution > Quantity: 100mL