Learn More
3M™ Safety-Walk™ Cushion Matting 5270E
This motor oil resistant mat, suitable for heavy foot traffic in dry areas, provides a comfortable surface for walking or standing.
Brand: 3M™ Safety-Walk™ 7100116723
This item is not returnable.
View return policy
Description
- Coiled cushion matting compresses evenly across the bottom of the shoe, reducing leg and back fatigue
- Textured top improves traction for safer footing
- Suitable for dry work areas such as assembly lines and production areas
- Supplied with edging to allow for wheeled traffic
- Withstands most industrial and cleaning chemicals
Specifications
Black | |
6.1 m | |
3 ft. |
20 ft. | |
Cushion Mat | |
0.91 m |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
3M™ Safety-Walk™ Cushion Matting 5270E > 6.1 m
Spot an opportunity for improvement?Share a Content Correction