Learn More
Thermo Scientific™ CyberScan TB 1000 Turbidity Meters
The CyberScan TB1000 series turbidity meters are designed to meet the stringent requirements of today's scientists.
£136.00 - £1953.58
Specifications
Automatic Range Selection | Yes |
---|---|
EPA Approved Methods | 180.1 |
ISO Accepted Methods | ISO 7027 |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11523755
|
Thermo Scientific™
01X265301 |
Tungsten filament lamp module (white light) for turbidity meter | N/A | N/A | N/A | |||||
10724247
|
Thermo Scientific™
01X265102 |
Calibration kit set for CyberScan TB 1000 - ECTBDIR1000 (0.02, 10.0, 1000 NTU) | N/A | N/A | N/A | |||||
11356340
|
Thermo Scientific™
ECTBDW100040 |
CyberScan TB 1000 white light turbidity bench meter (NTU) with calibration kit (ECTBDWCALKT) & 100/240 VAC power adapter | N/A | N/A | N/A | |||||
Description
The CyberScan TB1000 series turbidity meters are designed to meet the stringent requirements of today's scientists. TB1000 series meet the performance criteria specified by USEPA method 180.1 for NTU measurement and ISO 7027 (IR version). Ideal for beverage production, drinking water, waste water treatment, petrochemical and electroplating applications.
Features- multi-Point Push-Button Calibration (Up to 3 points) for optimum full-range accuracy of ± 2% full scale
- Auto-Ranging for fast response and best resolution (up to 0.01 NTU) over widemeasurement range
- Push-Button Calibration is hassle-free, quick and easy
- Auto-Alert Calibration prompts at user-defined interval to ensure regularly scheduled recalibration for ISO and GLP compliance
- RS232 Communication interface to download data to a printer or PC for report documentation
- Built-in Real-time Clock stamps stored data and calibration data with time & date -meets standards for GLP purpose
- Self-Diagnostic withmessage codes for easy troubleshooting
- Pour-Through Assemblymodule (optional) allows instantaneous sampling without having to transfer samples into cuvettes thus saving time.
Specifications
Yes | |
180.1 | |
ISO 7027 |