missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexane, 99+%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals Product Code.: 10348953

Cyclohexane, 99+%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

Product Code. 10348953
2.5 L, Glass Bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10348953

Brand: Thermo Scientific Acros 406020025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexaneShow MoreShow Less
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Absorbance 0.01 max. at 300nm, 0.01 max. at 400nm, 0.02 max. at 260nm, 0.03 max. at 250nm, 0.08 max. at 240nm, 0.2 max. at 230nm, 0.5 max. at 220nm, 1 max. at 210nm
Density 0.7700g/mL
Boiling Point 81.0°C
Quantity 2.5 L
Flash Point −18°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4250 to 1.4270
Assay Percent Range 99+%
Residue after Evaporation 0.002% max.
Specific Gravity 0.77
Linear Formula C6H12
Viscosity 0.94 mPa.s (20°C)
Beilstein 05, 20
Water 0.02% max. (Coulometric)
Merck Index 15, 2716
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Cyclohexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462886 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number406020025,406025000
Lot NumberA0443903 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number406020025,406025000
Lot NumberA0453030 Certificate TypeCertificate of Analysis Date10/05/2023 Catalog Number406020025,406025000
Lot NumberA0395443 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number406020025,406025000
Lot NumberA0432108 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog Number406020025,406025000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexane, 99+%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.