missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexane, 99+%, Thermo Scientific Chemicals Product Code.: 11403087

Cyclohexane, 99+%, Thermo Scientific Chemicals

Product Code. 11403087
1000 mL
Click to view available options
Quantity:
1000 mL
5000 mL
This item is not returnable. View return policy

Product Code. 11403087

Brand: Thermo Scientific Alfa Aesar A16070.AU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

Solubility
Miscible with ether, alcohol, acetone and olive oil. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexaneShow MoreShow Less
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.779
Boiling Point 80°C to 81°C
Quantity 1000 mL
Flash Point −18°C (−1°F)
Odor Pungent
Refractive Index 1.426
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
Solubility Information Miscible with ether,alcohol,acetone and olive oil. Immiscible with water.
Formula Weight 84.16
Percent Purity ≥99%
Chemical Name or Material Cyclohexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW03K035 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberA16070.0J,A16070.AU
Lot NumberT14J029 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberA16070.0J,A16070.AU
Lot Number10244924 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA16070.0J,A16070.AU
Lot NumberZ15I023 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog NumberA16070.0J,A16070.AU
Lot NumberW13I023 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog NumberA16070.0J,A16070.AU
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Cyclohexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexane, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.