missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, for HPLC, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Specifications
Specifications
Melting Point | 6.5°C |
Color | Colorless |
Boiling Point | 81°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 104mbar at 20°C |
Viscosity | 0.94 mPaS at 20°C |
Formula Weight | 84.15g/mol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2526771 | Certificate of Analysis | 13/02/2025 | C/8936/25RSS,C/8936/10RSS,C/8936/27SS,C/8936/PB17,C/8936/30RSS,C/8936/15,C/8936/17 |
Safety and Handling




Product Identifier
- Cyclohexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cyclohexane, for HPLC, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction