missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)Melibiose Monohydrate, Pure, Fisher Chemical
Click to view available options
10g, Plastic powder jar
Specifications
Specifications
CAS | 66009-10-7 |
Melting Point | 182°C |
Color | White |
Packaging | Plastic powder jar |
Molecular Formula | C12H22O11·H2O |
MDL Number | 198188 |
Quantity | 10 g |
Formula Weight | 360.32g/mol |
Physical Form | Powder |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Safety and Handling
Product Identifier
- D(+)-Melibiose monohydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
D(+)Melibiose Monohydrate, Pure, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction