missing translation for 'onlineSavingsMsg'
Learn More
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals Product Code.: 15299936

Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals

Product Code. 15299936
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 15299936

Brand: Thermo Scientific Acros 454582500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-02-6
Molecular Formula C10H30O5Si5
Molecular Weight (g/mol) 370.77
InChI Key XMSXQFUHVRWGNA-UHFFFAOYSA-N
Synonym decamethylcyclopentasiloxane, cyclopentasiloxane, decamethyl, cyclomethicone 5, dimethylsiloxane pentamer, dow corning 345 fluid, nuc silicone vs 7158, dow corning 345, silicon sf 1202, cyclic dimethylsiloxane pentamer, dekamethylcyklopentasiloxanShow MoreShow Less
PubChem CID 10913
IUPAC Name 2,2,4,4,6,6,8,8,10,10-decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)C

Specifications

Density 0.9590g/mL
Boiling Point 101.0°C (20.0 mmHg)
Flash Point 76°C
Assay Percent Range 96% min. (GC)
Quantity 250 g
Merck Index 15,2852
Specific Gravity 0.959
Solubility Information Solubility in water: insoluble
Viscosity 4 mPa.s (20°C)
Formula Weight 370.77
Percent Purity 97%
Chemical Name or Material Decamethylcyclopentasiloxane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454081 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number454580500,454582500
Lot NumberA0398121 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number454580500,454582500
Lot NumberA0394335 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number454580500,454582500
Lot NumberA0403595 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number454580500,454582500
Lot NumberA0404512 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number454580500,454582500
Product Identifier
  • Decamethylcyclopentasiloxane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.