missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Desiccator Sealing Ring
Desiccator Sealing Ring is a spare accessory for use with desiccators.
Brand: BRAND™ 65820
11.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sealing ring, CR
- For use with desiccator

Specifications
150 mm | |
Chloroprene Rubber | |
Black |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Desiccator Sealing Ring > Color: Black
Spot an opportunity for improvement?Share a Content Correction