Learn More
Invitrogen™ 293A Cell Line
Contains a stably integrated copy of the E1 gene that supplies the E1 proteins (E1a and E1b) required to generate recombinant adenovirus
Brand: Invitrogen™ R70507
491.40 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
507.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: 293A Cell Line
Description
A subclone of the 293 cell line and has a relatively flat morphology, it facilitates the initial production, amplification and titering of replication-incompetent adenovirus.
- Flat morphology of cells makes titering procedure easier and suitable for screening adenoviral constructs
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Constitutive Expression, Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery, Viral Vaccine Production

Specifications
1 vial of 1 x 107 cells. Store in liquid nitrogen. | |
Human | |
ViraPower™ |
293A | |
1 x 107 cells/mL |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.