Learn More
Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Gel
Bind and extract endotoxins (lipopolysaccharides or LPS) from antibody or protein samples using bulk resin or columns of immobilized Polymixin B ligand.
£235.00 - £4965.00
Specifications
Capacity (Metric) | 4000 to 6000 EU Binding per mL Gel |
---|---|
Content And Storage | Store at 4°C. |
Product Line | Detoxi-Gel™ |
Target | Endotoxin Removal |
Type | Endotoxin Removing Gel |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11866754
|
Thermo Scientific™
20339 |
10 mL |
£235.00
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11876754
|
Thermo Scientific™
20340 |
1000 mL |
£4965.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Detoxi-Gel Endotoxin Removing Gel and Columns provide a quick method to remove endotoxins (lipopolysaccharides or LPS) from samples.
Detoxi-Gel Resin is a beaded agarose support onto which polymixin B has been immobilized. This ligand binds to the lipid A portion of lipopolysaccharides (LPS), which are the dominant form of endotoxins that affect biological samples. Even low levels of endotoxins can be toxic to cells or organisms and must be removed before biological samples can be added to cells for various treatment experiments. Detoxi-Gel Resin provides a quick method for removal of endotoxins from samples. It is available in convenient pre-packed columns and two sizes of slurry for use in small-scale or large-scale processes.Highlights:
Affinity-based – uses immobilized polymixin B to remove endotoxins by binding to their lipid A domains
High capacity – 1mL of Detoxi-Gel Resin removes at least 9995 endotoxin units from a 5mL sample containing 10,000 EU of lipopolysaccharide (LPS)
Reusable – no loss in binding capacity even after 10 regenerations; requires activation and regeneration with 1% sodium deoxycholate (Part No. 89904)
Testing required – protein recovery is dependent upon protein type and concentration; empirical testing is required to validate the method for particular protein samples
Versatile – remove endotoxin from protein solutions, cell culture media and pharmacological components
Requires:
Sodium deoxycholate (Part No. 89904) for resin activation and regeneration.
Specifications
4000 to 6000 EU Binding per mL Gel | |
Detoxi-Gel™ | |
Endotoxin Removing Gel | |
Bottle |
Store at 4°C. | |
Endotoxin Removal | |
Polymixin B | |
Affinity, Agarose Resin |
Safety and Handling
- Detoxi-Gel Endotoxin Removing Gel
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.