missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Gel

Bind and extract endotoxins (lipopolysaccharides or LPS) from antibody or protein samples using bulk resin or columns of immobilized Polymixin B ligand.

Brand:  Thermo Scientific™ 20340

 View more versions of this product

Product Code. 11876754

  • £4965.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Detoxi-Gel Endotoxin Removing Gel and Columns provide a quick method to remove endotoxins (lipopolysaccharides or LPS) from samples.

Detoxi-Gel Resin is a beaded agarose support onto which polymixin B has been immobilized. This ligand binds to the lipid A portion of lipopolysaccharides (LPS), which are the dominant form of endotoxins that affect biological samples. Even low levels of endotoxins can be toxic to cells or organisms and must be removed before biological samples can be added to cells for various treatment experiments. Detoxi-Gel Resin provides a quick method for removal of endotoxins from samples. It is available in convenient pre-packed columns and two sizes of slurry for use in small-scale or large-scale processes.

Highlights:

  • Affinity-based – uses immobilized polymixin B to remove endotoxins by binding to their lipid A domains

  • High capacity – 1mL of Detoxi-Gel Resin removes at least 9995 endotoxin units from a 5mL sample containing 10,000 EU of lipopolysaccharide (LPS)

  • Reusable – no loss in binding capacity even after 10 regenerations; requires activation and regeneration with 1% sodium deoxycholate (Part No. 89904)

  • Testing required – protein recovery is dependent upon protein type and concentration; empirical testing is required to validate the method for particular protein samples

  • Versatile – remove endotoxin from protein solutions, cell culture media and pharmacological components

Requires:

Sodium deoxycholate (Part No. 89904) for resin activation and regeneration.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4000 to 6000 EU Binding per mL Gel
Detoxi-Gel™
Endotoxin Removal
Polymixin B
Bottle
Store at 4°C.
1000 mL
Endotoxin Removing Gel
4000 to 6000 EU per mL Gel
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberYF370194 Certificate TypeCertificate of Analysis Date15/07/2023 Catalog Number20340
Lot NumberXL364614 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number20340
Lot NumberXH362918 Certificate TypeCertificate of Analysis Date30/08/2022 Catalog Number20340
Lot NumberXG362520 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog Number20340
Lot NumberXD341383 Certificate TypeCertificate of Analysis Date21/04/2022 Catalog Number20340
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Detoxi-Gel™ Endotoxin Removing Gel > Detoxi-Gel Endotoxin Removing Gel; 1000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.