missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand: Fisherbrand™ 11958-13
323.29 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
Deuterium Lamp | |
400/185 nm | |
ABI 757 759 783A 785A 1000S 980 120A 130A |
UV | |
1 mm | |
1 Ea. |