missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Dewax and HIER Buffer L,M,H

Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.

Brand:  Epredia™ TA-999-DHBH


 View more versions of this product

Product Code. 15200715

  • £823.00 / 1 L
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dewax and HIER Buffer
10 x 100 mL
Buffer - High
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : titon-X100, EDTA, tetrasodium EDTA, tris(hydroxymethyl) aminomehtane, 2-methyl-3-isothiazolone, tet

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Dewax and HIER Buffer L,M,H > Buffer High; 10 x 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.