missing translation for 'onlineSavingsMsg'
Learn More

Di-Iso-Propyl Ether, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™

C6H14O, CAS Number-108-20-3, 2,2'-oxydipropane, 2-isopropoxypropane, bis isopropyl ether, diisopropylether, diisopropyl ether, diisopropyl oxide, ether, isopropyl, isopropylether, isopropyl ether, propane, 2,2'-oxybis, 2.5L, 68 deg.C, Colorless, 102.18g/mol, ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

£153.00 - £1645.00

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number 8880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10657022
View Documents Promotion Details
Fisher Chemical
D/5002/15
1 L Amber glass bottle
£153.00
1 L
Please to purchase this item. Need a web account? Register with us today!
10366180
View Documents Promotion Details
Fisher Chemical
D/5002/17
2.5 L Amber glass bottle
£199.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10699103
View Documents Promotion Details
Fisher Chemical
D/5002/21
5 L Metal can
£1060.00
5 L
Please to purchase this item. Need a web account? Register with us today!
10082923
View Documents Promotion Details
Fisher Chemical
D/5002/25
25 L Metal can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

C6H14O, CAS Number-108-20-3, 2,2'-oxydipropane, 2-isopropoxypropane, bis isopropyl ether, diisopropylether, diisopropyl ether, diisopropyl oxide, ether, isopropyl, isopropylether, isopropyl ether, propane, 2,2'-oxybis, 2.5L, 68 deg.C, Colorless, 102.18g/mol, ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Specifications

Chemical Identifiers

108-20-3
102.177
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
7914
CC(C)OC(C)C
C6H14O
8880
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
2-propan-2-yloxypropane

Specifications

108-20-3
Colorless
C6H14O
diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
CC(C)OC(C)C
102.177
1.42
Liquid
-85.5°C
68°C
8880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
2-propan-2-yloxypropane
7914
102.18g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Isopropyl ether

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

Supplemental information
  • MIXTURE LIST-Contains : Isopropyl ether, 2,6-Di-tert-butyl-p-cresol

SDS
Documents

Documents

Product Certifications