missing translation for 'onlineSavingsMsg'
Learn More
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals Product Code.: 10712251

Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10712251
25 g
Click to view available options
Quantity:
25 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 10712251

Brand: Thermo Scientific Acros 204110250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2893-78-9
Molecular Formula C3HCl2N3NaO3
Molecular Weight (g/mol) 220.949
MDL Number MFCD00006036
InChI Key UNWRHVZXVVTASG-UHFFFAOYSA-N
Synonym sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
PubChem CID 86657659
IUPAC Name 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
SMILES C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Specifications

Color White
pH 6 to 7 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Formula Weight 219.95
Percent Purity ≥97.5% (on dry substance)
Physical Form Crystals or Granules
Chemical Name or Material Dichloroisocyanuric acid sodium salt, 98%

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462884 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number204110250,204115000,204110025
Lot NumberA0462883 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number204110250,204115000,204110025
Lot NumberA0457656 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number204110250,204115000,204110025
Lot NumberA0410507 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number204110250,204115000,204110025
Lot NumberA0452127 Certificate TypeCertificate of Analysis Date24/04/2023 Catalog Number204110250,204115000,204110025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichloroisocyanuric acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.