missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals Product Code.: 11446336

Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11446336
500 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11446336

Brand: Thermo Scientific Alfa Aesar B23119.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-70-9
Molecular Formula C3H6Cl2Si
Molecular Weight (g/mol) 141.066
MDL Number MFCD00000492
InChI Key YLJJAVFOBDSYAN-UHFFFAOYSA-N
Synonym dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silaneShow MoreShow Less
PubChem CID 31299
IUPAC Name dichloro-ethenyl-methylsilane
SMILES C[Si](C=C)(Cl)Cl

Specifications

Boiling Point 92°C to 93°C
Melting Point -78°C
Quantity 500 g
UN Number UN2985
Beilstein 1740822
Flash Point 4°C (39°F)
Formula Weight 141.07
Percent Purity 97%
Odor Acrid
Refractive Index 1.43
Sensitivity Moisture sensitive
Density 1.087
Chemical Name or Material Dichloromethylvinylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240507 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberB23119.36,B23119.22
Lot Number10219023 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog NumberB23119.36,B23119.22
Lot Number10209628 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog NumberB23119.36,B23119.22
Lot NumberUSLF005105 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23119.36,B23119.22
Lot Number10136958 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23119.36,B23119.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.