missing translation for 'onlineSavingsMsg'
Learn More
Dichlorophenylborane, 97%, Thermo Scientific Chemicals Product Code.: 10083953

Dichlorophenylborane, 97%, Thermo Scientific Chemicals

Product Code. 10083953
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10083953

Brand: Thermo Scientific Acros 318550250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-51-8
Molecular Formula C6H5BCl2
Molecular Weight (g/mol) 158.82
MDL Number MFCD00013612
InChI Key NCQDQONETMHUMY-UHFFFAOYSA-N
Synonym phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boronShow MoreShow Less
PubChem CID 136678
IUPAC Name dichloro(phenyl)borane
SMILES B(C1=CC=CC=C1)(Cl)Cl

Specifications

Melting Point 7.0°C
Color Colorless
Density 1.2200g/mL
Boiling Point 175.0°C
Flash Point −14°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5BCl2
Refractive Index 1.5450 to 1.5470
Quantity 25 g
Specific Gravity 1.22
Formula Weight 158.82
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichlorophenylborane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455974 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number318550250,318550050
Lot NumberA0453673 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number318550250,318550050
Lot NumberA0399239 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number318550250,318550050
Lot NumberA0425927 Certificate TypeCertificate of Analysis Date10/08/2021 Catalog Number318550250,318550050
Lot NumberA0413643 Certificate TypeCertificate of Analysis Date08/12/2020 Catalog Number318550250,318550050
compliance-icons compliance-icons
Product Identifier
  • Dichlorophenylborane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dichlorophenylborane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichlorophenylborane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.