missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Diclofenac sodium, 98% Product Code.: 12317163

Thermo Scientific Chemicals Diclofenac sodium, 98%

Product Code. 12317163
10 g, Glass bottle
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 12317163

Brand: Thermo Scientific Chemicals 445250100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diclofenac sodium, CAS # 15307-79-6, is a nonsteroidal anti-inflammatory drug (NSAID) that inhibits prostaglandin synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Diclofenac is a is a nonsteroidal anti-inflammatory drug (NSAID) with anti-inflammatory and analgesic properties
  • Diclofenac has been shown to have anti-tumor capabilities through inhibition of MYC expression and glucose metabolism in models of melanoma and leukemia

Applications

  • Diclofenac sodium can be used as an antioxidant and free radical scavenger
  • It has been shown to have anti-bacterial activity against Listeria, E. coli, and M. tuberculosis partially through inhibition of bacterial DNA synthesis
  • It can be used for the study of metabolic activity as a CYP2C9 substrate
TRUSTED_SUSTAINABILITY

Specifications

CAS 15307-79-6
Melting Point 283.0°C to 285.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C14H10Cl2NNaO2
Quantity 10 g
Merck Index 15,3091
Synonym diclofenac sodium, sodium diclofenac, diclofenac sodium salt, voltaren, solaraze, anthraxiton, prophenatin, allvoran, batafil, delimon
Solubility Information Solubility in water: 50mg/mL
InChI Key KPHWPUGNDIVLNH-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)CC(=O)[O-])NC2=C(C=CC=C2Cl)Cl.[Na+]
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
Molecular Weight (g/mol) 318.13
PubChem CID 5018304
ChEBI CHEBI:4509
Formula Weight 318.13
Percent Purity 98%
Chemical Name or Material Diclofenac sodium
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458218 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number445250100
Lot NumberA0403708 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number445250100
Lot NumberA0409086 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number445250100
Lot NumberA0440854 Certificate TypeCertificate of Analysis Date06/04/2022 Catalog Number445250100
Lot NumberA0411795 Certificate TypeCertificate of Analysis Date04/03/2022 Catalog Number445250100
compliance-icons
Product Identifier
  • Diclofenac sodium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Diclofenac sodium, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.