missing translation for 'onlineSavingsMsg'
Learn More
Diethyl ether, 99+%, ACS reagent, anhydrous, stabilized with BHT, Thermo Scientific Chemicals Product Code.: 10351491

Diethyl ether, 99+%, ACS reagent, anhydrous, stabilized with BHT, Thermo Scientific Chemicals

Product Code. 10351491
2.5 kg
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10351491

Brand: Thermo Scientific Acros 410060025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.0°C
Density 0.7140g/mL
Boiling Point 34.6°C
Quantity 2.5 kg
Flash Point −45°C
Packaging Metal Drum
Refractive Index 1.3510 to 1.3530
Assay Percent Range 99% min. (GC)
Specific Gravity 0.714
Titratable Acid 0.0002 meq/g max
Linear Formula (CH3CH2)2O
Viscosity 0.2448 mPa.s (20°C)
Beilstein 01,314
Fieser 11,3762
Merck Index 15,3861
Solubility Information Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
Formula Weight 74.12
Percent Purity 99+%
Grade ACS Reagent
Chemical Name or Material Diethyl ether, Anhydrous, stabilized with BHT
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469315 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number410065000,410060025
Lot NumberA0423898 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number410065000,410060025
Lot NumberA0454454 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number410065000,410060025
Lot NumberA0450697 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number410065000,410060025
Lot NumberA0401781 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number410065000,410060025
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl ether, 99+%, ACS reagent, anhydrous, stabilized with BHT, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.