Learn More
Diethylene glycol diethyl ether, HPLC Grade, 99+%, Thermo Scientific Chemicals
Description
Diethylene glycol diethyl ether is used to study the activities of enzymes in aqueous organic mixtures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -44°C |
CAS | 112-36-7 |
Density | 0.909 g/mL |
Boiling Point | 188°C to 190°C |
Quantity | 250 mL |
Flash Point | 71°C (160°F) |
Physical Form | Liquid |
Refractive Index | 1.412 |
Molecular Formula | C8H18O3 |
MDL Number | MFCD00009254 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N28J024 | Certificate of Analysis | 09/05/2023 | 043464.AK |
G30S040 | Certificate of Analysis | 07/12/2021 | 043464.AK |
Z17F056 | Certificate of Analysis | 07/12/2021 | 043464.AK |
E24S014 | Certificate of Analysis | 07/12/2021 | 043464.AK |
G24L20 | Certificate of Analysis | 07/12/2021 | 043464.AK |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.