missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals Product Code.: 10507031

Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals

Product Code. 10507031
5 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10507031

Brand: Thermo Scientific Acros 338160050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54731-72-5
Molecular Formula C4H11O4P
Molecular Weight (g/mol) 154.10
MDL Number MFCD00043376
InChI Key TZPPDWDHNIMTDQ-UHFFFAOYSA-N
Synonym dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-olShow MoreShow Less
PubChem CID 2733833
IUPAC Name 2-dimethoxyphosphorylethanol
SMILES COP(=O)(CCO)OC

Specifications

Color Colorless to Yellow
Density 1.2470g/mL
Boiling Point 98.0°C to 104.0°C (0.1 mmHg)
Flash Point 192°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2P(O)(CH2)2OH
Refractive Index 1.4430 to 1.4460
Quantity 5 mL
Beilstein 04, IV, 3584
Specific Gravity 1.247
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 154.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dimethyl 2-hydroxyethylphosphonate, 95%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0400406 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number338160250,338160050
Lot NumberA0419147 Certificate TypeCertificate of Analysis Date07/11/2022 Catalog Number338160250,338160050
Lot NumberA0447092 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog Number338160250,338160050
compliance-icons
Product Identifier
  • Dimethyl 2-hydroxyethylphosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.