missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl sulfate, 99+%, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10512318

Dimethyl sulfate, 99+%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10512318
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10512318

Brand: Thermo Scientific Acros 430831000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-78-1
Molecular Formula C2H6O4S
Molecular Weight (g/mol) 126.13
MDL Number MFCD00008416
InChI Key VAYGXNSJCAHWJZ-UHFFFAOYSA-N
Synonym dimethyl sulphate, sulfuric acid, dimethyl ester, dimethyl monosulfate, dimethylsulfate, dimethylsulfat, sulfate dimethylique, sulfuric acid dimethyl ester, dimethylsulfaat, dimetilsolfato, dms methyl sulfateShow MoreShow Less
PubChem CID 6497
ChEBI CHEBI:59050
IUPAC Name dimethyl sulfate
SMILES COS(=O)(=O)OC

Specifications

Melting Point -32.0°C
Density 1.3300g/mL
Boiling Point 188.0°C
Flash Point 83°C
Assay Percent Range 99+%
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)2SO4
Refractive Index 1.3860 to 1.3880
Quantity 100 mL
Merck Index 15,328
Specific Gravity 1.33
Solubility Information Solubility in water: 2.8g/100mL (18°C). Other solubilities: hydrolysis rapidly at 18°C or above in water,soluble in ehter,dioxane,acetone and aromatic,hydrocarbons,sparingly soluble in carbon disulfide and,aliphatic hydrocarbons
Viscosity 1.81 mPa.s (20°C)
Formula Weight 126.13
Percent Purity 99+%
Chemical Name or Material Dimethyl sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466877 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number430830010,430831000
Lot NumberA0462037 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number430830010,430831000
Lot NumberA0460169 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number430830010,430831000
Lot NumberA0458231 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog Number430830010,430831000
Lot NumberA0453918 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number430830010,430831000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethyl sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • Multiple Manufacturers
    Use code 24096 to receive this promotion.
    Promo Code: 24096

    Expires2025-05-16

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 21/04/2025 and 16/05/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.

    Terms & Conditions Learn More

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl sulfate, 99+%, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.