missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ≥99.9%, Honeywell Riedel-de Haën™

ACS Reagent,≥99.9%

£56.13 - £1384.82

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O
View More Specs

Products 8
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15645630
View Documents
Honeywell-Riedel-de Haen
472301-100ML
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15685630
View Documents
Honeywell-Riedel-de Haen
472301-500ML
500 mL
£113.08
500mL
Please to purchase this item. Need a web account? Register with us today!
15655630
View Documents Promotion Details
Honeywell-Riedel-de Haen
1 L
£157.50
1 L
Please to purchase this item. Need a web account? Register with us today!
15665630
View Documents
Honeywell-Riedel-de Haen
472301-2.5L
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15605640
View Documents
Honeywell-Riedel-de Haen
472301-6X500ML
6 x 500 mL
£587.31
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15658410
View Documents
Honeywell-Riedel-de Haen
472301-4L
4 L
£552.99
4 L
Please to purchase this item. Need a web account? Register with us today!
15695630
View Documents
Honeywell-Riedel-de Haen
472301-6X1L
6 x 1 L
£573.62
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15675630
View Documents
Honeywell-Riedel-de Haen
472301-4X4L
4 x 4 L
£1384.82
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CS(C)=O
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262

Specifications

67-68-5
1.10g/mL
87°C (188.6°F)
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(C)=O
78.13
CHEBI:28262
≥99.9%
Dimethyl sulfoxide
Colorless
42%, 63°F
573°F
(CH3)2SO
0.42 mmHg (20°C)
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methanesulfinylmethane
679
78.13
ACS Reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : 200-664-3

SDS
Documents

Documents

Product Certifications