missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals
£13.00 - £50.90
Chemical Identifiers
CAS | 120-61-6 |
---|---|
Molecular Formula | C10H10O4 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00008440 |
InChI Key | WOZVHXUHUFLZGK-UHFFFAOYSA-N |
Synonym | dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate |
PubChem CID | 8441 |
SMILES | COC(=O)C1=CC=C(C=C1)C(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10255710
|
Thermo Scientific Acros
180580250 |
25 g | Plastic bottle |
£13.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10225760
|
Thermo Scientific Acros
180585000 |
500 g | Plastic bottle |
£17.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10012420
|
Thermo Scientific Acros
180580010 |
1 kg | Plastic bottle |
£25.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10722611
|
Thermo Scientific Acros
180580025 |
2.5 kg | Plastic bottle |
£50.90
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-61-6 | |
194.19 | |
WOZVHXUHUFLZGK-UHFFFAOYSA-N | |
8441 |
C10H10O4 | |
MFCD00008440 | |
dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate | |
COC(=O)C1=CC=C(C=C1)C(=O)OC |
Specifications
0.03mg KOH/g max. | |
140°C to 143°C | |
288°C | |
Authentic | |
C10H10O4 | |
09, 843 | |
dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate | |
285.0 °C | |
COC(=O)C1=CC=C(C=C1)C(=O)OC | |
194.19 | |
194.19 | |
Flakes or Pellets |
120-61-6 | |
White | |
154°C | |
98.5% min. (GC) | |
MFCD00008440 | |
15, 9305 | |
Solubility in water: 0.036g/L (20°C). Other solubilities: soluble in ether and hot alcohol | |
WOZVHXUHUFLZGK-UHFFFAOYSA-N | |
dimethyl benzene-1,4-dicarboxylate | |
8441 | |
99% | |
Dimethyl terephthalate, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 204-411-8
missing translation for 'rtecsNumber' : WZ1225000
missing translation for 'tsca' : TSCA
RUO – Research Use Only