missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals Product Code.: 10255710

Dimethyl terephthalate, 99%, Thermo Scientific Chemicals

Product Code. 10255710
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10255710

Brand: Thermo Scientific Acros 180580250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-61-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008440
InChI Key WOZVHXUHUFLZGK-UHFFFAOYSA-N
Synonym dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoateShow MoreShow Less
PubChem CID 8441
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Acidity 0.03mg KOH/g max.
Melting Point 140°C to 143°C
Color White
Boiling Point 288°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 09, 843
Merck Index 15, 9305
Solubility Information Solubility in water: 0.036g/L (20°C). Other solubilities: soluble in ether and hot alcohol
Sublimation Point 285.0 °C
IUPAC Name dimethyl benzene-1,4-dicarboxylate
Formula Weight 194.19
Percent Purity 99%
Physical Form Flakes or Pellets
Chemical Name or Material Dimethyl terephthalate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467086 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number180585000,180580010,180580250,180580025
Lot NumberA0455022 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number180585000,180580010,180580250,180580025
Lot NumberA0373482 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number180585000,180580010,180580250,180580025
Lot NumberA0397509 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number180585000,180580010,180580250,180580025
Lot NumberA0395849 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number180585000,180580010,180580250,180580025
Product Identifier
  • Dimethyl terephthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl terephthalate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.