Learn More
Brady™ Directional Arrow Tape
Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
£41.50 - £164.00
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
---|---|
Description | Directional Arrow Tape |
Finish | Gloss |
Format | Tape |
Material | Polyester |
Product Code | Brand | Dimensions (LxW) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Color | Price | Quantity & Availability | ||||
12747926
|
Brady™
275131 |
33 m x 100 mm | Black on Gray |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12707936
|
Brady™
275128 |
33 m x 100 mm | Black on Orange |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12797936
|
Brady™
275129 |
33 m x 100 mm | Black on White |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12747946
|
Brady™
275124 |
33 m x 100 mm | Black on Yellow |
£163.00
1 roll |
In Stock
Log in to see stock. |
||||
12757916
|
Brady™
275133 |
33 m x 100 mm | White on Black |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12787916
|
Brady™
275127 |
33 m x 100 mm | White on Blue |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12717926
|
Brady™
275130 |
33 m x 100 mm | White on Brown |
£164.00
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12767926
|
Brady™
275125 |
33 m x 100 mm | White on Green |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12737936
|
Brady™
275132 |
33 m x 100 mm | White on Purple |
£164.00
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12767936
|
Brady™
275126 |
33 m x 100 mm | White on Red |
£164.00
1 roll |
In Stock
Log in to see stock. |
||||
12797926
|
Brady™
275107 |
33 m x 25 mm | Black on Gray |
£44.05
1 roll |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
12707946
|
Brady™
275105 |
33 m x 25 mm | Black on White |
£44.05
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12797916
|
Brady™
275103 |
33 m x 25 mm | White on Blue |
£44.05
1 roll |
In Stock
Log in to see stock. |
||||
12759664
|
Brady™
275110 |
33 m x 25 mm | Black on Ochre |
£41.50
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12777926
|
Brady™
275101 |
33 m x 25 mm | White on Green |
£45.85
1 roll |
In Stock
Log in to see stock. |
||||
12717936
|
Brady™
275104 |
33 m x 25 mm | Black on Orange |
£44.05
1 roll |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
12777936
|
Brady™
275102 |
33 m x 25 mm | White on Red |
£44.05
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12767916
|
Brady™
275109 |
33 m x 25 mm | White on Black |
£44.05
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12727926
|
Brady™
275106 |
33 m x 25 mm | White on Brown |
£44.05
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12747936
|
Brady™
275108 |
33 m x 25 mm | White on Purple |
£44.05
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12727946
|
Brady™
275100 |
33 m x 25 mm | Black on Yellow |
£44.05
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12757926
|
Brady™
275119 |
33 m x 50 mm | Black on Gray |
£72.80
1 roll |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
12727936
|
Brady™
275116 |
33 m x 50 mm | Black on Orange |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12717946
|
Brady™
275117 |
33 m x 50 mm | Black on White |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12757946
|
Brady™
275112 |
33 m x 50 mm | Black on Yellow |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12769664
|
Brady™
275122 |
33 m x 50 mm | Black on Ochre |
£68.75
1 roll |
In Stock
Log in to see stock. |
||||
12777916
|
Brady™
275121 |
33 m x 50 mm | White on Black |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12707926
|
Brady™
275115 |
33 m x 50 mm | White on Blue |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12737926
|
Brady™
275118 |
33 m x 50 mm | White on Brown |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12787926
|
Brady™
275113 |
33 m x 50 mm | White on Green |
£75.55
1 roll |
In Stock
Log in to see stock. |
||||
12757936
|
Brady™
275120 |
33 m x 50 mm | White on Purple |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
12787936
|
Brady™
275114 |
33 m x 50 mm | White on Red |
£72.80
1 roll |
In Stock
Log in to see stock. |
||||
Description
- Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
Gloss | |
Polyester | |
Pipe and valve marking | |
1300 Pk. | |
0.0039 in. | |
108.27 ft. |
Directional Arrow Tape | |
Tape | |
Pipe Marker | |
1300 Markers/Roll | |
-40°C to +110°C | |
0.099 mm | |
33 m |