missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Homocysteinethiolactone hydrochloride, 99% Product Code.: 10103111

Thermo Scientific Chemicals DL-Homocysteinethiolactone hydrochloride, 99%

Product Code. 10103111
100 g, Glass bottle
Click to view available options
Quantity:
1 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10103111

Brand: Thermo Scientific Chemicals 120831000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6038-19-3
MDL Number MFCD00012724
InChI Key ZSEGSUBKDDEALH-UHFFFAOYSA-N
Synonym dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochlorideShow MoreShow Less
PubChem CID 110753
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES C1CSC(=O)C1N.Cl

Specifications

Melting Point 199.0°C to 203.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H7NOS·HCl
Quantity 100 g
Merck Index 15, 4772
Solubility Information Solubility in water: 740.5g/L (20 c)
Sublimation Point >140°C
Molecular Weight (g/mol) 153.63
Formula Weight 153.63
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Homocysteinethiolactone hydrochloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461816 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog Number120830250,120831000
Lot NumberA0421618 Certificate TypeCertificate of Analysis Date06/02/2023 Catalog Number120830250,120830010,120831000
Lot NumberA0392605 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number120830250,120830010,120831000
Lot NumberA0407720 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number120830250,120830010,120831000
Lot NumberA0416049 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number120830250,120830010,120831000
Product Identifier
  • DL-Homocysteinethiolactone hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Homocysteinethiolactone hydrochloride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.