missing translation for 'onlineSavingsMsg'
Learn More
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals Product Code.: 10745771

DL-Mandelic acid, 99+%, Thermo Scientific Chemicals

Product Code. 10745771
250 g, Plastic bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10745771

Brand: Thermo Scientific Acros 125312500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key IWYDHOAUDWTVEP-UHFFFAOYSA-N
Synonym mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acidShow MoreShow Less
PubChem CID 1292
ChEBI CHEBI:35825
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 117°C to 121°C
Color White
Assay Percent Range 99+%
Linear Formula C6H5CH(OH)CO2H
Formula Weight 152.15
Percent Purity ≥99.0% (Acid)
Quantity 250 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material DL-Mandelic acid, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398912 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number12531018,125310050,125312500,125310010,125311000
Lot NumberA0393328 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number12531018,125310050,125312500,125310010,125311000
Lot NumberA0404971 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number12531018,125310050,125312500,125310010,125311000
Lot NumberA0416997 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number12531018,125310050,125312500,125310010,125311000
Lot NumberA0407657 Certificate TypeCertificate of Analysis Date29/08/2022 Catalog Number12531018,125310050,125312500,125310010,125311000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.