missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ dNTPs Mix (10mm each)

Improves PCR efficiency, lowers denaturation temperatures and increases template lengths. MP Biomedicals dNTPs Mix (10mm each) is a ready to use solution with guaranteed purification of dNTPs (>99% pure) in order to achieve sensitive and consistent res

£92.10 - £362.00

Specifications

pH 7
For Use With (Application) Standard PCR, Real Time PCR (qPCR), RT-PCR
Concentration 10 mM
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11457120
View Documents
MP Biomedicals™
11NTPMX100
5 μmol
£92.10
5µmol
Please to purchase this item. Need a web account? Register with us today!
12846895
View Documents
MP Biomedicals™
11NTPMX105
5 x 5 μmol
£362.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manufacturing process guarantees that the dNTPs are highly purified, as contaminants can hinder PCR and result in a disappointing product yield. This product achieves sensitive and consistent results with specially prepared deoxynucleotides at a concentration of 10mm.

It is suitable for use in a range of molecular biology applications, including: PCR, real-time PCR, high fidelity and long PCR, RT-PCR, cDNA synthesis, and DNA sequencing and labeling procedures.

Specifications

Specifications

7
Standard PCR, Real Time PCR (qPCR), RT-PCR
10 mM
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ dNTPs Mix (10mm each)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.