missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals

£22.31 - £148.74

Chemical Identifiers

CAS 151-21-3, 7757-82-6, 7647-14-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10761691
View Documents
Thermo Scientific Acros
226141000
100 g Plastic Bottle
£22.31
100g
Please to purchase this item. Need a web account? Register with us today!
10002810
View Documents
Thermo Scientific Acros
226145000
500 g Plastic Bottle
£44.92
500g
Please to purchase this item. Need a web account? Register with us today!
11964561
View Documents
Thermo Scientific Acros
226140025
2.5 kg Plastic drum
£148.74
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

151-21-3, 7757-82-6, 7647-14-5
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
sodium dodecyl sulfate
C12H25NaO4S
MFCD00036175
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
CHEBI:8984
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

White to Yellow
Powder or Crystals
>150°C
85%
CH3(CH2)11OSO3Na
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
3423265
288.38
Pure
151-21-3, 7757-82-6, 7647-14-5
206.0°C
Authentic
C12H25NaO4S
MFCD00036175
Solubility in water: 130g/L (20°C).
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
85%
Dodecyl sulfate, sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Danger

Product Identifier
  • Dodecyl sulfate sodium salt

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only