missing translation for 'onlineSavingsMsg'
Learn More
Dodecylamine, 98%, Thermo Scientific Chemicals Product Code.: 10122960

Dodecylamine, 98%, Thermo Scientific Chemicals

Product Code. 10122960
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10122960

Brand: Thermo Scientific Acros 117661000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-22-1
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.36
MDL Number MFCD00008154
InChI Key JRBPAEWTRLWTQC-UHFFFAOYSA-N
Synonym dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4Show MoreShow Less
PubChem CID 13583
IUPAC Name dodecan-1-amine
SMILES CCCCCCCCCCCCN

Specifications

Melting Point 28.0°C to 32.0°C
Color White
Density 0.8
Boiling Point 247.0°C to 249.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Alkyl chain distribution C12) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11NH2
Quantity 100 g
Beilstein 04, 200
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, isopropylalcohol, ethanol,, ether
Formula Weight 185.35
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Dodecylamine, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452080 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number117665000,117660010,117661000
Lot NumberA0444084 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number117665000,117660010,117661000
Lot NumberA0432570 Certificate TypeCertificate of Analysis Date11/10/2021 Catalog Number117665000,117660010,117661000
Lot NumberA0353425 Certificate TypeCertificate of Analysis Date28/04/2021 Catalog Number117665000,117660010,117661000
Lot NumberA0424273 Certificate TypeCertificate of Analysis Date09/11/2020 Catalog Number117665000,117660010,117661000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecylamine
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dodecylamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.