Learn More
Doxorubicin Hydrochloride, Fisher BioReagents
£42.15 - £204.00
Chemical Identifiers
CAS | 25316-40-9 |
---|---|
Molecular Formula | C27H30ClNO11 |
Molecular Weight (g/mol) | 579.98 |
MDL Number | MFCD00077757,MFCD00077757,MFCD00941448 |
InChI Key | MWWSFMDVAYGXBV-FGBJBKNOSA-N |
Synonym | doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs |
PubChem CID | 129626538 |
IUPAC Name | (7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride |
SMILES | [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10512955
|
Fisher BioReagents
BP2516-1 |
1 mg | Amber Glass | ||||||
11413074
|
Fisher BioReagents
BP2516-5 |
5 mg | Amber Glass | ||||||
10133293
|
Fisher BioReagents
BP2516-10 |
10 mg | Amber Glass | ||||||
10767945
|
Fisher BioReagents
BP2516-50 |
50 mg | Amber Glass | ||||||
Description
Doxorubicin Hydrochloride
Doxorubicin HCL is a chemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. It blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis.Chemical Identifiers
25316-40-9 | |
579.98 | |
MWWSFMDVAYGXBV-FGBJBKNOSA-N | |
129626538 | |
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O |
C27H30ClNO11 | |
MFCD00077757,MFCD00077757,MFCD00941448 | |
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs | |
(7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride |
Specifications
25316-40-9 | |
216°C | |
(by HPLC): ≥95% | |
MFCD00077757,MFCD00077757,MFCD00941448 | |
doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs | |
MWWSFMDVAYGXBV-FGBJBKNOSA-N | |
(7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride | |
129626538 | |
Powder or Solid |
216°C | |
Orange | |
C27H30ClNO11 | |
15, 3487 | |
Soluble in water | |
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O | |
579.98 | |
580 | |
Doxorubicin Hydrochloride |
Safety and Handling
WARNING!
Emergency Overview
May cause cancer. Risk of serious damage to eyes. Irritating to respiratory system and skin. May cause heritable genetic damage. Possible risk of impaired fertility. Possible risk of harm to the unborn child. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.
NFPA
Health:2
Flammability:1
Instability:0
Recommended Storage : 2° to 8°C
- Doxorubicin hydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.