Learn More
Dräger™ Short-Term Detector Tubes - Nitrogen Fumes
For making spot measurments for various gases and vapors
Brand:  Dräger™ CH27701
 View more versions of this product
Product Code. 12896663
Description
Description
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Specifications
Nitrous Fumes 100/c
100 to 5000ppm
10
50°F to 86°F
30mg/L
5/1+4
1.5 min./18 sec.
10°C to 30°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Dräger™ Short-Term Detector Tubes - Nitrogen Fumes > Nitrous fumes 100/c; Measuring range: 100 to 5000ppm