missing translation for 'onlineSavingsMsg'
Learn More

Drummond™ Microcap™ Disposable Micropipets

Fast, safe, clean, and accurate way to transfer liquids in micro quantities

Brand:  Drummond™ 07950-05

27.14 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11888661

  • £31.05 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Precision-bore glass capillary tubes
  • Disposable microcaps
  • With spotting holder and bulb dispenser

Specifications

Specifications

0.0039 in.
0.09 mm
Glass
Spotting holder and bulb dispenser
0.5 μL
Micropipet
0.021 in.
0.53 mm
For handling dangerous materials, fast, safe, clean and accurate way to transfer liquids in micro quantities
1.25 in.
32 mm
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Drummond™ Microcap™ Disposable Micropipets > 0.5uL0.5μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.