missing translation for 'onlineSavingsMsg'
Learn More

Manutan Duo clad back shelving extension bay, six shelves, 2450mm x 450mm

Brand:  Manutan A338203

Encompass_Preferred

Product Code. 12835108

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Extend your shelving bay with this robust double skin shelving. Compatible with all Stormor shelving. BioCote antibacterial coating to prevent the growth of bacteria and fungi, reducing the risk of cross-contamination from surfaces. Cladded uprights provide strength and stability with slim profile shelves and frames to minimise wasted space. Components are finished in hard-wearing grey paint RAL7035 with 25mm increments to allow shelves to be adjusted easily. Supplied flat-packed with all necessary fixings and assembly instructions for easy self-assembly.

  • Six shelves
  • Shelf capacity UDL: 70kg
  • Dimensions (H x W x D): 2450mm x 1000mm x 450mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Manutan Duo clad back shelving extension bay, six shelves, 2450mm x 450mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.