Learn More
DWK Life Sciences DURAN™ Bell Jar, with glass knob top
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).
Brand:  DWK Life Sciences 244605902
 View more versions of this product
Product Code. 12739666
  • £244.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
  • The button makes it easy to raise the bell jar
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
185 mm
Vacuum applications; Tests in vacuum chambers
Autoclavable
Borosilicate Glass (3.3)
1/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Bell Jar, with glass knob top > Diameter 185 mm