Learn More
Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Dysprosium Aceate has specialized uses in ceramics, glass, phosphors, lasers and Dysprosium Metal halide lamp. High purity of Dysprosium Aceate is used in electronics industry as an antireflection coating in photoelectric devices. Dysprosium is used in conjunction with vanadium and other elements, in making laser materials and commercial lighting. Dysprosium and its compounds are highly susceptible to magnetization, they are employed in various data-storage applications, such as in hard disks. It is also used in dosimeters for measuring ionizing radiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 120°C (decomposition) |
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
Solubility Information | Soluble in water,moderately soluble in strong mineral acids. |
Formula Weight | 411.68 (339.62 Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Odor | Odorless |
Assay | (REO) |
Chemical Name or Material | Dysprosium(III) acetate tetrahydrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y14K013 | Certificate of Analysis | 19/11/2024 | 014595.14,014595.22 |
Z11J068 | Certificate of Analysis | 08/01/2024 | 014595.14,014595.22 |
X28H014 | Certificate of Analysis | 17/11/2022 | 014595.14,014595.22 |
S10F017 | Certificate of Analysis | 08/12/2021 | 014595.14,014595.22 |
S10F014 | Certificate of Analysis | 08/12/2021 | 014595.14,014595.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.