missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11312548

Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11312548
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11312548

Brand: Thermo Scientific Alfa Aesar 014595.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dysprosium Aceate has specialized uses in ceramics, glass, phosphors, lasers and Dysprosium Metal halide lamp. High purity of Dysprosium Aceate is used in electronics industry as an antireflection coating in photoelectric devices. Dysprosium is used in conjunction with vanadium and other elements, in making laser materials and commercial lighting. Dysprosium and its compounds are highly susceptible to magnetization, they are employed in various data-storage applications, such as in hard disks. It is also used in dosimeters for measuring ionizing radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15280-55-4
Molecular Formula C6H17DyO10
Molecular Weight (g/mol) 411.69
MDL Number MFCD00013033
InChI Key SIYZPSYZNCQFDV-UHFFFAOYSA-K
IUPAC Name dysprosium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Dy+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point 120°C (decomposition)
Physical Form Crystalline Aggregates
Quantity 25 g
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 411.68 (339.62 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Assay (REO)
Chemical Name or Material Dysprosium(III) acetate tetrahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY14K013 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number014595.14,014595.22
Lot NumberZ11J068 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number014595.14,014595.22
Lot NumberX28H014 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number014595.14,014595.22
Lot NumberS10F017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014595.14,014595.22
Lot NumberS10F014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014595.14,014595.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.