missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta), Thermo Scientific Chemicals Product Code.: 11323808

Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11323808
25 mm
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
This item is not returnable. View return policy

Product Code. 11323808

Brand: Thermo Scientific Alfa Aesar 040285.DH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.27g/mm, Cast rod, diameter is approximate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Gray to Silver
Physical Form Cast Rod
Quantity 25 mm
Assay Percent Range 99.7% (Metals basis excluding Ta)
Odor Odorless
Appearance Gray to silver rod
Chemical Name or Material Dysprosium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS25K002 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number040285.DH,040285.HF,040285.DM,040285.DJ
Lot NumberC13M30 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040285.DH,040285.HF,040285.DM,040285.DJ
Lot NumberF24W048 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040285.DH,040285.HF,040285.DM,040285.DJ
Lot NumberR06A019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040285.DH,040285.HF,040285.DM,040285.DJ
Lot NumberZ14D032 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040285.DH,040285.HF,040285.DM,040285.DJ

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.