Learn More
Contec™ EasySat Mop
Designed for the cleanroom with a built-in 52 ounce solution tank that completely eliminates the need for a bucket and wringer. Contec™ EasySat Mop is an effective and quick solution to your cleaning challenges.
Brand: Contec™ BLMOPE
This item is not returnable.
View return policy
Description
- Hydrophilic polyester/clean ester, laser cut sealed edge foam material.
- Translucent tank allows user to see solution
- Fully autoclavable for critical arease
- Easy to use
- Compatible with a wide range of solvents
Specifications
Bucketless Mop | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
126810-220727 | Certificate of Quality | 12/01/2023 | BLMOPE |
126422-220705 | Certificate of Quality | 08/10/2022 | BLMOPE |
210321 | Certificate of Compliance | 29/07/2021 | BLMOPE |
Product Content Correction
Product Title
Contec™ EasySat Mop > Dimensions (L x W): 55 x 46 cm
Spot an opportunity for improvement?Share a Content Correction