missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Elite pH Spear Pocket Testers and Replacement Sensors

Flexible on-the-go pH testing using durable Elite spear pocket testers with replaceable sensors for a variety of applications include food, beverage, agriculture and more!

Brand:  Thermo Scientific™ ELITEPHSPEARKIT

 View more versions of this product

Product Code. 15934022

  • £279.70 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.

Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite pH Spear model to measure pH using a spear-shaped sensor design for your application. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.

  • Flexible testing option
  • Measure pH
  • Large back-lit graphic display
  • Uses 4 x AAA alkaline batteries
  • Average 250 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
  • One-year warranty for tester
  • Six-month warranty for replacement sensors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.02
250 hr. (average)
pH
CE, FCC
Graphic Dot Matrix
pH
0 to 80 ppt
0.0°C to 60.0°C
5 to 85 %, non-condensing
Manual input
5°C to 45°C (ambient)
0.1°C, 0.1°F
Yes
4 x AAA Alkaline
1 to 3
Elite pH spear pocket tester for measuring pH with a spear-tip sensor
IP 67
-1 to 15
0.01
Elite pH spear tester with batteries, lanyard, and pH buffer
Stability icon on display
Yes with manual temperature input
32.0°F to 140.0°F
1 Year for Tester
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers