Learn More
Thermo Scientific™ Elite pH Spear Pocket Testers and Replacement Sensors
Flexible on-the-go pH testing using durable Elite spear pocket testers with replaceable sensors for a variety of applications include food, beverage, agriculture and more!
£255.56 - £279.66
Specifications
Accuracy (pH) | ±0.02 |
---|---|
Measurement Modes | pH |
Range (pH) | -1 to 15 |
Resolution (pH) | 0.01 |
Temperature Range (Metric) | 0.0°C to 60.0°C |
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | ||||
15934022
|
Thermo Scientific™
ELITEPHSPEARKIT |
Elite pH spear pocket tester for measuring pH with a spear-tip sensor | Elite pH spear tester with batteries, lanyard, and pH buffer |
£279.66
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15924022
|
Eutech
ELITEPHSPEAR |
Elite pH spear pocket tester for measuring pH | Elite pH spear tester with batteries and lanyard |
£255.56
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.
Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite pH Spear model to measure pH using a spear-shaped sensor design for your application. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
- Flexible testing option
- Measure pH
- Large back-lit graphic display
- Uses 4 x AAA alkaline batteries
- Average 250 hours of battery life
- Wide leak-proof cap serves as stable base
- Replaceable sensors
- One-year warranty for tester
- Six-month warranty for replacement sensors
Specifications
±0.02 | |
-1 to 15 | |
0.0°C to 60.0°C | |
5°C to 45°C (ambient) | |
0.1°C, 0.1°F |
pH | |
0.01 | |
Manual input | |
32.0°F to 140.0°F |